Order Entry
Canada
Orders LinkContactUsLinkComponent
RIPA Buffer, Abcam

RIPA Buffer, Abcam

RIPA Buffer, 100 ml
Catalog # ABCA_AB288006-100M
New Product
Supplier:   Abcam
Supplier #:  AB288006-100ML
RIPA Buffer, Abcam
Catalog # ABCA_AB288006-100M
Supplier:  Abcam
New Product
CAS Number:  
RIPA Buffer, Abcam

RIPA Buffer, 100 ml

CAS Number:  
Price
$257.00
Log in for availability
Each (100ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A ready-to-use buffer useful for lysing cells and tissues for radioimmunoprecipitation assay.

  • Constituents: 1% Alcohol Ethoxylate, 0.877% Sodium chloride, 0.788% Tris HCl, 0.5% Sodium deoxycholate, 0.1% Sodium lauryl sulfate

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    RIPA Buffer
  • Size:
    100 ml
  • Cat. No.:
    ABCA_AB288006-100M
  • Storage temperature:
    Store at -20°C.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.