Order Entry
Canada
Orders LinkContactUsLinkComponent
Extraction Buffer 5X PTR, Abcam

Extraction Buffer 5X PTR, Abcam

Extraction Buffer 5X PTR, 50 ml
Catalog # ABCA_AB193970-50ML
New Product
Supplier:   Abcam
Supplier #:  AB193970-50ML
Extraction Buffer 5X PTR, Abcam
Catalog # ABCA_AB193970-50ML
Supplier:  Abcam
New Product
CAS Number:  
Extraction Buffer 5X PTR, Abcam

Extraction Buffer 5X PTR, 50 ml

CAS Number:  
Price
$113.00
Log in for availability
Each (50ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Extraction buffer 5X PTR is formulated for preparing cell or tissue extracts for use in immunoassays, with added phosphatase inhibitors and protease inhibitor aprotinin. - Cited in over 20 publications - Phosphatase and protease inhibitors – contains inhibitors to preserve sample integrity - Easy to use – dilute to 1X with distilled water before use, or use with extraction enhancer buffer 50X for enhanced extraction - Compatible with a variety of sample types

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Extraction Buffer 5X PTR
  • Size:
    50 ml
  • Cat. No.:
    ABCA_AB193970-50ML
  • Storage temperature:
    Store at +4°C.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.