ANTIBODY ANTI-LAMB3 OTI3A2 A322318 100UG
Catalog # 77970-886
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-LAMB3 OTI3A2 A322318 100UG
Catalog # 77970-886
Supplier: ANTIBODIES.COM LLC
Supplier Number: A322318-100
New Product
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Water, deionized VWR®
Supplier: VWR
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Single-Use Needles, BD Medical
Supplier: BD
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Enzymes and Separation Accessories, Omega Bio-Tek
Supplier: Omega Bio-Tek
PhosVer® 3 Phosphate Reagent Powder Pillows, Hach
Supplier: Hach