ANTIBODY HUMAN POLYCLONAL UBE2M 100UL
Catalog # 77968-569
Supplier: Sino Biological
New Product
ANTIBODY HUMAN POLYCLONAL UBE2M 100UL
Catalog # 77968-569
Supplier: Sino Biological
Supplier Number: 101017-T32-100
New Product
Frequently Bought Together
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Quality Economy Brand Luer Slip Syringes, Air-Tite
Supplier: Air-Tite
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Supplier: VWR International
Sodium chloride for biotechnology
Supplier: VWR
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Cryoscope Standards, Advanced Instruments
Supplier: Advanced Instruments
Tissue Marking Dyes, Azer Scientific
Supplier: Azer Scientific
Corning® Donor Horse Serum, US Sourced, Corning
Supplier: Corning
Corning® rat collagen I (from tail tendon)
Supplier: Corning
BBL™ SFP Agar Base, BD
Supplier: BD
Transport Vials, Caplugs
Supplier: Caplugs
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex