ELISA KIT HORSE IL-1RA A4228 48 TEST
Catalog # 77966-626
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT HORSE IL-1RA A4228 48 TEST
Catalog # 77966-626
Supplier: ANTIBODIES.COM LLC
Supplier Number: A4228-48
New Product
Frequently Bought Together
BD Angiocath™ IV Catheters, BD™
Supplier: BD
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Calcium chloride, anhydrous ≥96% ACS
Supplier: VWR
Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®
Supplier: MilliporeSigma
Vesphene® III st Disinfectant Cleaners
Supplier: Steris
Hose Clamps, Adjustable, Stainless Steel
Supplier: VWR International
Diisopropyl azodicarboxylate 94%
Supplier: Thermo Scientific Chemicals
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
Cutting Mats, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
E.Z.N.A.® Plasmid Mini Kit I, Omega Bio-tek®
Supplier: Omega Bio-Tek
Ricinoleic acid ≥80.0%
Supplier: TCI America
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
BsoBI
Supplier: New England Biolabs (NEB)
SporAmpule® Liquid Biological Indicator
Supplier: HARDY DIAGNOSTICS CA