ELISA KIT CANINE IL-1RA A4227 48 TEST
Catalog # 77967-214
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT CANINE IL-1RA A4227 48 TEST
Catalog # 77967-214
Supplier: ANTIBODIES.COM LLC
Supplier Number: A4227-48
New Product
Frequently Bought Together
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Sodium chloride, VWR® USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
MiniCollect® Serum and Plasma Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Phenolphthalein 1% in alcohol analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
N,N-Dimethylformamide ≥99.8% (by GC), GR ACS, Supelco®
Supplier: MilliporeSigma
PfuUltra II Fusion HS DNA Polymerase, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA