BUFFER STAINING FUSION 600ML
Catalog # MSPP-240198
Supplier: AKOYA BIOSCIENCES, INC MS
BUFFER STAINING FUSION 600ML
Catalog # MSPP-240198
Supplier: AKOYA BIOSCIENCES, INC MS
Supplier Number: 240198
Frequently Bought Together
Sodium hydroxide 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
Supplier: Micro Essential
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
Preserved Starfish
Supplier: Avantor
Sodium molybdate(VI) dihydrate 99.5-103.0% ACS
Supplier: Thermo Scientific Chemicals
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Immunoreagents
Sodium perchlorate, anhydrous 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
Molybdate Reagent Powder Pillows for HR Silica, Hach
Supplier: Hach
Tetramethoxysilane 98%
Supplier: Thermo Scientific Chemicals
Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Cypermethrin Single-Component Organic Standard, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
Test Kits, COD, TNT plus®
Supplier: Hach
Iron(III) chloride hexahydrate 97%
Supplier: Thermo Scientific Chemicals