ANTIBODY ANTI-NF2 A16638 100UL
77964-193
New Product
Frequently Bought Together
VWR® Sterile 70% Isopropanol
: VWR International
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
: VWR International
SYRINGE STERILE WITHOUT NEEDLE 10ML
: AMX INDUSTRIAL TRADE MX
Orion™, All-in-One™ pH Buffers Kit, Thermo Scientific
: Thermo Fisher Scientific
VWR® Reagent alcohol
: VWR International
TapeStation Systems Parts and Accessories
: AGILENT TECHNOLOGIES (GENOMICS) CA
Sol-Vet® Standard Insulin Syringes (Veterinary Care), Sol M
: SOL-MILLENNIUM CANADA, INC. CA
Corn Meal Agar, CRITERION™, Hardy Diagnostics
: HARDY DIAGNOSTICS CA
High Sensitivity RNA ScreenTape Analysis
: AGILENT TECHNOLOGIES (GENOMICS) CA
N,N-Dimethylformamide ≥99.9%, OmniSolv®, Supelco®
: MilliporeSigma
Cornstarch
: Ward's Science
HISTOSETTE® II Cassettes in QuickLoad ™ Sleeve, Simport
: Simport Scientific
Accessories for Gas Regulators
: VWR International
NEEDLE HYPODERMC TERUMO 26GX1/2IN CS1000
: Cardinal Health
Prepared Blood Agar Media
: Culture Media & Supplies
Ascorbic Acid
: Ward's Science
Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
: Simport Scientific