ELISA KIT AMYLOID BETA 1-40 A1802 48TEST
77962-827
New Product
Frequently Bought Together
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
: VWR International
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
: VWR International
Ethanol denatured, anhydrous
: AVANTOR PERFORMANCE MATERIAL LLC
di-Potassium hydrogen phosphate 98.0-100.5% (dried basis) USP, Multi-Compendial endotoxin-free, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC
Water USP, Ph. Eur., purified
: Ricca Chemical
Hexacarbonylmolybdenum 98%
: Thermo Scientific Chemicals
Acetylacetone 99%
: Thermo Scientific Chemicals
Ascorbic Acid
: Ward's Science
Calcium Nitrate 4-Hydrate
: Ward's Science
QUICK-LOAD® Taq 2X Master Mix, New England Biolabs
: New England Biolabs (NEB)