ELISA KIT HUMAN PDP1/PDP A1997 48 TEST
Catalog # 77963-177
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT HUMAN PDP1/PDP A1997 48 TEST
Catalog # 77963-177
Supplier: ANTIBODIES.COM LLC
Supplier Number: A1997-48
New Product
Frequently Bought Together
Starswabs II® Specimen Collection and Transport Systems
Supplier: Starplex
COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
Sodium acetate, anhydrous ACS
Supplier: VWR
VACUETTE® EDTA Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Paper Tapes, DUKAL™ Corporation
Supplier: Dukal
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
2-Deoxy-D-glucose 98%
Supplier: Thermo Scientific Chemicals
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Reagent Sets for Colorimeters, Hach
Supplier: Hach
ES 7X™ Concentrate, MP Biomedicals
Supplier: MP Biomedicals
L(-)-Glutathione (reduced form) 97%
Supplier: Thermo Scientific Chemicals
Cryoscope Standards, Advanced Instruments
Supplier: Advanced Instruments
Sodium hydroxide standardised solution 0.1 N in aqueous solution
Supplier: Thermo Scientific Chemicals
Buffer Solution
Supplier: Ward's Science
Skeletal Muscle Cell Growth Medium, PromoCell
Supplier: Promocell
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton