Order Entry
Canada
Orders LinkContactUsLinkComponent
Mouse eNOS ELISA Kit

Mouse eNOS ELISA Kit

Mouse eNOS ELISA Kit, 96 tests
Catalog # 77962-850
New Product
Supplier:   Antibodies.com
Supplier #:  A1813-96
Mouse eNOS ELISA Kit
Catalog # 77962-850
Supplier:  Antibodies.com
New Product
CAS Number:  
Mouse eNOS ELISA Kit

Mouse eNOS ELISA Kit, 96 tests

CAS Number:  
Price
$1,362.77
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Mouse eNOS ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse eNOS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids.

Mouse eNOS ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse eNOS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. An antibody specific for eNOS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the eNOS present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-eNOS Antibody, which binds the captured eNOS present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-colored product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of eNOS captured in each well. The concentration of eNOS can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Specifications

Specifications

  • Description:
    Mouse eNOS ELISA Kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids
  • Detection range:
    1.56 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cat. No.:
    77962-850
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0.67 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    eNOS
  • Detection limit:
    Intra-assay: CV <10%, Inter-assay: CV <12%
  • Regulatory status:
    RUO

Specifications