ELISA KIT HUMAN ALAD A4577 48 TEST
Catalog # 77961-443
Supplier: Antibodies.com
New Product
ELISA KIT HUMAN ALAD A4577 48 TEST
Catalog # 77961-443
Supplier: Antibodies.com
Supplier Number: A4577-48
New Product
Frequently Bought Together
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Masterflex® I/P® Spooled Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
n-Dodecane ≥99.0%
Supplier: TCI America
LabKlenz™ 250 Acid Detergents
Supplier: Steris
InLab® Conductivity Probes, Mettler Toledo
Supplier: Mettler Toledo
VACUUBRAND® ME 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Supplier: Brandtech
Ligation Sequencing Kits
Supplier: Oxford Nanopore Technologies
Gastight® 1800 Series Syringes, Hamilton
Supplier: Hamilton
Silver nitrate 0.500 N (N/2) in aqueous solution
Supplier: Ricca Chemical
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International