ELISA KIT HUMAN CD24 A4613 96 TEST
Catalog # 77961-502
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT HUMAN CD24 A4613 96 TEST
Catalog # 77961-502
Supplier: ANTIBODIES.COM LLC
Supplier Number: A4613-96
New Product
Frequently Bought Together
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Heptane (mixture of isomers) ACS
Supplier: INNOVA CHEMICALS CA
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Ammonium chloride ≥99.5% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
X-tourn™ Tourniquets
Supplier: Heathrow Scientific
Prinzing® Ball Valve Lockouts, Brady Worldwide®
Supplier: Brady Worldwide
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
Difco™ Raka-Ray No.3 Medium, BD
Supplier: BD
qScript™ cDNA SuperMix, QuantaBio
Supplier: Quantabio
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA