ANTIBODY ANTI-TAK1 A90981 100UL
Catalog # 77960-301
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-TAK1 A90981 100UL
Catalog # 77960-301
Supplier: ANTIBODIES.COM LLC
Supplier Number: A90981-100
New Product
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
Calcium chloride dihydrate 99.0-107.0% USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
VWR® Blood Bank Saline Solutions
Supplier: VWR International
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
VWR® Specimen Containers, PP with PE Caps
Supplier: VWR International
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Supplier: VWR International
MEDIUM SDA BBL SETTLING PLATE STERILE PK
Supplier: BD
VACUUM PUMP VACUUBRAND ME1C
Supplier: Brandtech
HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Supplier: Ansell Canada
Cerium(IV) sulfate solution 0.100 N (N/10) in sulphuric acid 2N
Supplier: Ricca Chemical
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
Blunt Needle Luer Stub Adaptors, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
EDTA 0.800 M, cartridge for digital titrator
Supplier: Hach