ELISA KIT MOUSE PSP A7324 96 TEST
Catalog # 77956-953
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT MOUSE PSP A7324 96 TEST
Catalog # 77956-953
Supplier: ANTIBODIES.COM LLC
Supplier Number: A7324-96
New Product
Frequently Bought Together
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
GelRed® & GelGreen® Nucleic Acid Gel Stains, Biotium
Supplier: Biotium
VWR® Disposable Petri Dishes
Supplier: VWR International
Sterile IPA 70% Spray, ITW Texwipe
Supplier: ITW Texwipe
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
SYRINGE STERILE WITHOUT NEEDLE 10ML
Supplier: AMX INDUSTRIAL TRADE MX
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: HARDY DIAGNOSTICS CA
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning