ELISA KIT HUMAN PSP A7230 96 TEST
Catalog # 77956-781
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT HUMAN PSP A7230 96 TEST
Catalog # 77956-781
Supplier: ANTIBODIES.COM LLC
Supplier Number: A7230-96
New Product
Frequently Bought Together
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Ammonium sulfate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
Reagent Sets for Colorimeters, Hach
Supplier: Hach
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA