ELISA SERINE PROTEASE 33 A6471 96TEST
Catalog # 77955-331
Supplier: ANTIBODIES.COM LLC
New Product
ELISA SERINE PROTEASE 33 A6471 96TEST
Catalog # 77955-331
Supplier: ANTIBODIES.COM LLC
Supplier Number: A6471-96
New Product
Frequently Bought Together
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Puritan® Standard Cotton Tipped Applicators, Wood Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
Orion™, All-in-One™ pH Buffers Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetone ACS for production, VWR Chemicals BDH®
Supplier: VWR International
VWR® Reagent alcohol
Supplier: VWR International
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
4,4'-Dimethylbenzophenone ≥99.0%
Supplier: TCI America
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Cardboard Freezer Boxes and Dividers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Fluorescamine
Supplier: TCI America
COMPLEMENT GUINEA PIG NON-STERILE 10ML
Supplier: Rockland Immunochemical
Feather™ Scalpel Handle with Blade, Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences