ELISA KIT MOUSE PRP A6205 48 TEST
Catalog # 77950-795
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT MOUSE PRP A6205 48 TEST
Catalog # 77950-795
Supplier: ANTIBODIES.COM LLC
Supplier Number: A6205-48
New Product
Frequently Bought Together
VWR® Blood Bank Saline Solutions
Supplier: VWR International
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Flanged Plug Caps, Tiered, 13 mm, Globe Scientific
Supplier: Globe Scientific
Ammonium sulfate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
Dimethyl sulfoxide ACS
Supplier: VWR
VWR® Reagent alcohol
Supplier: VWR International
Eveready® Alkaline Batteries, Bulbtronics
Supplier: Bulbtronics
BETA-GARD™ Waste Containers, IBI Scientific
Supplier: IBI Scientific
1-Cetylpyridinium chloride monohydrate 98%
Supplier: Thermo Scientific Chemicals
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
ClearSteel, Stainless Steel Cleaner and Polish Spray, Mortech
Supplier: Mortech Manufacturing
Adjustable Well Plate Stand, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Reinforced Clostridial Medium, BD Diagnostics
Supplier: BD
Histosette® I Tissue Processing/Embedding Cassettes, Simport
Supplier: Simport Scientific
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences