ELISA KIT RAT IL-17F A6421 48 TEST
Catalog # 77955-232
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT RAT IL-17F A6421 48 TEST
Catalog # 77955-232
Supplier: ANTIBODIES.COM LLC
Supplier Number: A6421-48
New Product
Frequently Bought Together
High Vacuum Grease, DuPont
Supplier: Liveo
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
VWR® Specimen Containers, PP with PE Caps
Supplier: VWR International
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Leakbuster™ Specimen Containers
Supplier: Starplex
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
CL17 DPD Indicator, Hach
Supplier: Hach
BBL™ SFP Agar Base, BD
Supplier: BD
BPMO ≥99% (by 1H-NMR), high purity
Supplier: Enzo Life Sciences
Anti-APP Mouse Monoclonal Antibody [clone: 9C4/Amyloid]
Supplier: Biolegend
Chloroform ≥99.8% stabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
BUFFER SOLN TOT CHLORINE 473ML
Supplier: Hach