ELISA KIT MOUSE RYR A5270 96 TEST
Catalog # 77949-971
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT MOUSE RYR A5270 96 TEST
Catalog # 77949-971
Supplier: ANTIBODIES.COM LLC
Supplier Number: A5270-96
New Product
Frequently Bought Together
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Single-Use Needles, BD Medical
Supplier: BD
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Puritan® Tongue Depressors, Puritan Medical Products
Supplier: Puritan Medical Products
BathBrite™ Cleaner
Supplier: Decon Labs
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Prinzing® Ball Valve Lockouts, Brady Worldwide®
Supplier: Brady Worldwide
Bromothymol blue 0.04% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
MOPS Minimal Medium Kit, Teknova
Supplier: Teknova
Carbon Steel Surgical Blades, Non Sterile, Cincinnati Surigcal
Supplier: CINCINNATI SURGICAL CO.
COMPLEMENT GUINEA PIG NON-STERILE 10ML
Supplier: Rockland Immunochemical