ELISA KIT MOUSE IL-2RA A7474 96 TEST
Catalog # 77945-734
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT MOUSE IL-2RA A7474 96 TEST
Catalog # 77945-734
Supplier: ANTIBODIES.COM LLC
Supplier Number: A7474-96
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Quality Economy Brand Luer Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
D-(+)-Maltose monohydrate, Reagent Grade
Supplier: VWR
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Supplier: Ricca Chemical
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Corning® Donor Horse Serum, US Sourced, Corning
Supplier: Corning
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Tissue Marking Dyes, Azer Scientific
Supplier: Azer Scientific