ANTIBODY HUMAN CTSV RABBIT PAB 100UL.
Catalog # 77940-977
Supplier: Sino Biological
New Product
ANTIBODY HUMAN CTSV RABBIT PAB 100UL.
Catalog # 77940-977
Supplier: Sino Biological
Supplier Number: 10093-T24-100
New Product
Frequently Bought Together
Bulk Syringes, Nonsterile, BD Medical
Supplier: BD
Leakbuster™ Specimen Containers
Supplier: Starplex
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
Acetonitrile ≥99.8% for HPLC, Supelco®
Supplier: MilliporeSigma
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Single-Use Needles, BD Medical
Supplier: BD
Hydrazinium sulfate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Low-EDTA TE (1X), pH 8.0, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
Blunt Tip Needles with Dispensing Tips, Air-Tite
Supplier: Air-Tite
Magnesium
Supplier: Ward's Science
Swann Morton Surgical Blade Handle #5, Ergonomic, Cincinnati Surgical
Supplier: CINCINNATI SURGICAL CO.
BUFFER SOLN TOT CHLORINE 473ML
Supplier: Hach
ACK Lysing Buffer, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.