NEUROENDOCRINE REGULATORY PEPTIDE 100UG
Catalog # MSPP-076-96
Supplier: Phoenix Pharmaceuticals
NEUROENDOCRINE REGULATORY PEPTIDE 100UG
Catalog # MSPP-076-96
Supplier: Phoenix Pharmaceuticals
Supplier Number: 076-96
Frequently Bought Together
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Sulfuric acid 95.0-98.0% ACS
Supplier: Anachemia
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Blood Bank Saline Solutions
Supplier: VWR International
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
Acetone ACS for production, VWR Chemicals BDH®
Supplier: VWR International
TapeStation Systems Parts and Accessories
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
MacConkey Broth Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
Solvent Resistant Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
AccuStart™ II PCR Genotyping Kit, Quantabio
Supplier: Quantabio
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Ligation Sequencing Kits
Supplier: Oxford Nanopore Technologies
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Toluene ACS
Supplier: INNOVA CHEMICALS CA