Orders LinkContactUsLinkComponent
Safe-Store Slide File Systems | Avantor
Safe-Store Slide File Systems
8-drawer Safe-Store slide file system
Catalog # 77908-046
New Product
Supplier:   VWR International
Safe-Store Slide File Systems
Catalog # 77908-046
Supplier:  VWR International
New Product
Safe-Store Slide File Systems
8-drawer Safe-Store slide file system
Price
$355.52
Log in for availability
Case of 10
Product Details & Documents
Economical interlocking corrugated storage for slides and cassettes.
  • Tabs allow stacking
    Available as a standard 4-drawer file, or larger capacity 8-drawer file
When used as a 4-drawer file, holds approximately 2000 slides. When used as an 8-drawer file holds approximately 500 tissue blocks.
Specifications
  • Description:
    8-drawer Safe-Store slide file system