Orders LinkContactUsLinkComponent
PathFlow Supa Mega Cassettes
PathFlow Supa Mega Cassettes
PathFlow Supa Mega cassettes, Blue
Catalog # 77908-006
New Product
Supplier:   StatLab
Supplier #:  EAG-0106-02A
PathFlow Supa Mega Cassettes
Catalog # 77908-006
Supplier:  StatLab
New Product
PathFlow Supa Mega Cassettes
PathFlow Supa Mega cassettes, Blue
Price
$619.95
Log in for availability
Box of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Supa Mega Cassette is particularly useful for prostate, brain, breast tissue, and eye. As a result, this cassette has been widely adopted by clinical, research, and toxicology laboratories.
  • Better reagent flow and improved turnaround times
    Proven writing surface
    Large color range available
The Supa Mega Cassette design offers better sectioning quality, with the innovative hex design ensuring a greater adhesion of the paraffin block to the cassette. This eliminates the possibility of the embedded specimen being separated from the cassette when sectioning fibrous specimens.
Specifications
  • Color:
    Blue
  • Description:
    PathFlow Supa Mega cassettes
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.