Slide Boxes, HDPE
Product Details & Documents
Blue plastic slide boxes made from durable HDPE .
- Hinged lid
- Foam interior cushioning
- Equipped with locking mechanism
- 12 boxes in each case
Specifications
- Color:Blue
- Description:100-place slide box
- Material:HDPE
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)