PathFlow Supa Mega Base Mold
PathFlow Supa Mega base mold, 36×36×10 mm
Catalog # 77908-056
Supplier: StatLab
Supplier #: GBC-3614-05A
Jump to:
Product Details & Documents
PathFlow stainless steel Supa Mega base molds.
- Reusable Stops wax leakage Stackable design for efficient storage Stainless steel for a lifetime of use
Specifications
- Description:PathFlow Supa Mega base mold
- Dimensions:36×36×10 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.