Wallcur® PRACTI-Labeled Vials
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Prelabeled Vials For Convenience
- Contains No Medication
- Suitable For Most Simulator Applications
Each of these vials is already labeled in the same way as their real counterparts are! Available in a wide variety of simulated labels, each contains all the information a normal label does as well as additional warnings for training purposes. Vials are also bar coded making your pharma training easier for inventory management. Materials containing only Distilled water are generally suitable for simulator use. Those containing other materials are not recommended for use in simulators. All of these factors make these vials ideal for skills training for nursing, EMS and pharm tech programs.
Have students practice all the skills needed for proper drug selection, dosing and administration. These Vials help teach:
Reading and interpreting dosage requirements
Calculating commonly ordered antihistamine drug dosages
Small vial pressure changes
Small needle syringe techniques
Injection administration skills
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Documents
Product Purchase Authorization Wallcur Version 3.pdf
Additional Product Details
Specifications
- Description:Practi-amiodarone (150 mg/3 ml) tint vial
- Includes:Distilled water