LACTO-N-NEOTETRAOSE 10MG
Catalog # 77889-053
Supplier: AFG Bioscience
New Product
LACTO-N-NEOTETRAOSE 10MG
Catalog # 77889-053
Supplier: AFG Bioscience
Supplier Number: C79028-10MG
New Product
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Supplier: Ricca Chemical
Glucose Tolerance Drinks, Azer Scientific
Supplier: Azer Scientific
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Optical Lens Cleaner, FoxWest Sales
Supplier: Decon Labs
Isoamyl alcohol ≥98.5% ACS
Supplier: Thermo Scientific Chemicals
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
Succinimide 98+%
Supplier: Thermo Scientific Chemicals
Tri-Corn Beakers, Plastic, Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Sodium alginate, high viscosity
Supplier: Thermo Scientific Chemicals
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC