-GLCNAC-PEG3-THIOL 25MG
Catalog # 77888-901
Supplier: AFG Bioscience
New Product
-GLCNAC-PEG3-THIOL 25MG
Catalog # 77888-901
Supplier: AFG Bioscience
Supplier Number: C53012-25MG
New Product
Frequently Bought Together
Alconox® Powdered Precision Cleaners
Supplier: Alconox
Ethyl Alcohol Denatured
Supplier: Ward's Science
Puritan® Standard Cotton Tipped Applicators, Wood Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
VWR® Sterile 70% Isopropanol
Supplier: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Exel International® Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
1-Naphthoyl chloride 98%
Supplier: Thermo Scientific Chemicals
VWR® Grade 707 Blotting Paper, Grade 707
Supplier: VWR International
Chloroform-[D₁] (99.8% D) (Isotopic) + 0.03% v/v TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
VACUUM PUMP VACUUBRAND ME1C
Supplier: Brandtech
InLab® Conductivity Probes, Mettler Toledo
Supplier: Mettler Toledo
Pepsin
Supplier: Ward's Science
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals