3-AZDPRPYLΑ-DMANP 1 2 ΑD-MANP 1 2 Α 10MG
Catalog # 77888-821
Supplier: AFG Bioscience
New Product
3-AZDPRPYLΑ-DMANP 1 2 ΑD-MANP 1 2 Α 10MG
Catalog # 77888-821
Supplier: AFG Bioscience
Supplier Number: C51786-10MG
New Product
Frequently Bought Together
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Clear-Rite 3™ Clearant, Epredia
Supplier: Thermo Scientific
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Tissue Marking Dyes (TMD™), TBS®, General Data
Supplier: General Data Healthcare
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
HistoGel™ Specimen Processing Gel, Epredia
Supplier: Thermo Scientific
Succinimide 98+%
Supplier: Thermo Scientific Chemicals
Sodium perchlorate monohydrate 97+%
Supplier: Thermo Scientific Chemicals
E 5 LIQ MACH DISHWSH UN 4L
Supplier: Ostrem Chemical
Simple Nutrients Standard, NSI
Supplier: SPEX CERTIPREP LLC
Benzil 98%
Supplier: Thermo Scientific Chemicals
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics