Manβ1-4GlcNAc ≥95%
Glass vial, 10 mg
Catalog # 77889-036
Supplier: AFG Bioscience
: C74115-10MG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
: Store at 0 to 8 °C.
Synonyms: β-1,4-mannosylNAcetylglucosamine
- CAS Number:856224-94-7
- Formula:C₁₄H₂₅NO₁₁
- Molecular Weight:383.35 g/mol
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Glass vial
- Size:10 mg



