1-AZIDO1-DXY-Β-D-GLCPYRANURON 250MG
Catalog # 77888-804
Supplier: AFG Bioscience
New Product
1-AZIDO1-DXY-Β-D-GLCPYRANURON 250MG
Catalog # 77888-804
Supplier: AFG Bioscience
Supplier Number: C51700-250MG
New Product
Frequently Bought Together
AccuStart™ II GelTrack PCR SuperMix, QuantaBio
Supplier: Quantabio
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Phenol ≥99.0% saturated with buffer pH 4.5
Supplier: VWR
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
VWR® Lab Pen black ink
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane ≥99%
Supplier: Thermo Scientific Chemicals
Lysing Matrix B, MP Biomedicals
Supplier: MP Biomedicals
Sulfide 1 Reagent, 100 ml, Hach
Supplier: Hach
MORPHOLINE STANDARD, 1000PPM
Supplier: Labchem