2-AZIDOETHYL N-ACTYL--D-MNOSAMINE 25MG
Catalog # 77888-709
Supplier: AFG Bioscience
New Product
2-AZIDOETHYL N-ACTYL--D-MNOSAMINE 25MG
Catalog # 77888-709
Supplier: AFG Bioscience
Supplier Number: C51508-25MG
New Product
Frequently Bought Together
Citric acid 10% in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Leakbuster™ Specimen Containers
Supplier: Starplex
Bulk Syringes, Nonsterile, BD Medical
Supplier: BD
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Pocket Colorimeter II Test Kits
Supplier: Hach
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Sodium salicylate 99%
Supplier: Thermo Scientific Chemicals
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
QUICK-LOAD® Taq 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC