N-ACETYLMURAMIC ACID 250MG
Catalog # 77888-703
Supplier: AFG Bioscience
New Product
N-ACETYLMURAMIC ACID 250MG
Catalog # 77888-703
Supplier: AFG Bioscience
Supplier Number: C51086-250MG
New Product
Frequently Bought Together
Rubber Laboratory Apron
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Puritan® Standard Cotton Tipped Applicators, Wood Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
CHEMets® Visual Kit, Hydrogen Peroxide Test Kit, CHEMetrics
Supplier: CHEMetrics
THREADLOCKER 222 BOTTLE 50ML
Supplier: Loctite
TUBE K2EDTA MINICOLLECT 0.25/0.5ML PK500
Supplier: Greiner Bio-One
VWR® Reagent alcohol
Supplier: VWR International
ES 7X™ Concentrate, MP Biomedicals
Supplier: MP Biomedicals
Gastight® 1700 Series Syringes, Hamilton
Supplier: Hamilton
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Free and Total Chlorine Test Strips, Hach
Supplier: Hach
Magnesium Chloride Hexahydrate
Supplier: Ward's Science
HISTOSETTE® II Cassettes in QuickLoad ™ Sleeve, Simport
Supplier: Simport Scientific
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
Calcein AM fluorescent dye, Corning®
Supplier: Corning
Flow Cell Wash Kits
Supplier: Oxford Nanopore Technologies