P-NITROPHENYL-BETA-D-RIBOFURANOSD 100MG
Catalog # 77888-920
Supplier: AFG Bioscience
New Product
P-NITROPHENYL-BETA-D-RIBOFURANOSD 100MG
Catalog # 77888-920
Supplier: AFG Bioscience
Supplier Number: C53510-100MG
New Product
Frequently Bought Together
BD Microtainer® Tubes, BD Diagnostics
Supplier: BD
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Phenol ≥99.0% saturated with buffer pH 4.5
Supplier: VWR
Simulated ABO and Rh Blood Typing Lab Activity
Supplier: Avantor
VWR® Specimen Containers, PP with PE Caps
Supplier: VWR International
D-(+)-Maltose monohydrate, Reagent Grade
Supplier: VWR
Chloroform-[D₁] (99.8% D) (Isotopic) + 0.03% v/v TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
Accessories for eVol® Digital Analytical Syringe, SGE
Supplier: Trajan Scientific and Medical
Reagent Sets for Colorimeters, Hach
Supplier: Hach
Chloroform ≥99.9% stabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Single-Use Needles, BD Medical
Supplier: BD
D-(+)-Glucose solution 45%, Corning®
Supplier: Corning