4-FLUORO-N-ACETYLGLUCOSAMINE 5MG
Catalog # 77888-875
Supplier: AFG Bioscience
New Product
4-FLUORO-N-ACETYLGLUCOSAMINE 5MG
Catalog # 77888-875
Supplier: AFG Bioscience
Supplier Number: C52508-5MG
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
D-(+)-Sucrose, Ultrapure DNase-, RNase-free
Supplier: VWR
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
CHEMets® Visual Kit, Hydrogen Peroxide Test Kit, CHEMetrics
Supplier: CHEMetrics
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Phenols, CHEMets Visual Kit, CHEMetrics
Supplier: CHEMetrics
HENKE-JECT® 3-Part Disposable Syringes, Air-Tite
Supplier: Air-Tite
Nunc-Immuno™ MicroWell™ 96-Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Oakton® ORP Electrodes, Cole-Parmer
Supplier: Antyila Scientific
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
Cerium(IV) sulfate solution 0.100 N (N/10) in sulphuric acid 2N
Supplier: Ricca Chemical
Calcein AM fluorescent dye, Corning®
Supplier: Corning
Dri-Contrad® Powder Detergent
Supplier: Decon Labs
EDTA 0.800 M, cartridge for digital titrator
Supplier: Hach