-MAN-PEG3-ACID PERACETATE 25MG
Catalog # 77888-861
Supplier: AFG Bioscience
New Product
-MAN-PEG3-ACID PERACETATE 25MG
Catalog # 77888-861
Supplier: AFG Bioscience
Supplier Number: C52223-25MG
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Contrad® 70 Liquid Detergent
Supplier: Decon Labs
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Sterile IPA 70% Spray, ITW Texwipe
Supplier: ITW Texwipe
Single-Use Needles, BD Medical
Supplier: BD
VWR® Disposable Petri Dishes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VACUETTE® Heparin Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: HARDY DIAGNOSTICS CA
Reagent Sets for Colorimeters, Hach
Supplier: Hach