-MAN-PEG3-ACID PERACETATE 100MG
Catalog # 77888-860
Supplier: AFG Bioscience
New Product
-MAN-PEG3-ACID PERACETATE 100MG
Catalog # 77888-860
Supplier: AFG Bioscience
Supplier Number: C52223-100MG
New Product
Frequently Bought Together
MacConkey Agar, BD
Supplier: BD
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Calcium chloride dihydrate 99.0-107.0% USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Toluene ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Ascorbic Acid
Supplier: Ward's Science
CytoFLEX Sheath Fluid, Beckman Coulter
Supplier: Beckman Coulter
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA