N-ACTYL-9-AZIDO9-DEOXY-NEURAMINICACD 5MG
Catalog # 77888-775
Supplier: AFG Bioscience
New Product
N-ACTYL-9-AZIDO9-DEOXY-NEURAMINICACD 5MG
Catalog # 77888-775
Supplier: AFG Bioscience
Supplier Number: C51589-5MG
New Product
Frequently Bought Together
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
n-Hexane ≥95%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Single-Use Needles, BD Medical
Supplier: BD
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Water for HPLC, Supelco®
Supplier: MilliporeSigma
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Reagent Sets for Colorimeters, Hach
Supplier: Hach
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Supplier: VWR International
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC