2-ACTYLAMINO-6AZIDO-26-DIDXY-D-GLC 250MG
Catalog # 77888-736
Supplier: AFG Bioscience
New Product
2-ACTYLAMINO-6AZIDO-26-DIDXY-D-GLC 250MG
Catalog # 77888-736
Supplier: AFG Bioscience
Supplier Number: C51540-250MG
New Product
Frequently Bought Together
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
Silver nitrate 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
Acetone ≥99.5% ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Methanol ≥99.8% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Supplier: VWR International
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Water, deionized VWR®
Supplier: VWR
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Pierce Zeba™ Desalt Spin Columns, Thermo Scientific
Supplier: Thermo Scientific
Corning® DMEM/Ham's F-12 50/50 Mix Products, Corning
Supplier: Corning
qScript™ cDNA SuperMix, QuantaBio
Supplier: Quantabio