BZY 2 4-DI-O-BENZOYL-Α-DMANNOPYRNS 500MG
Catalog # 77888-673
Supplier: AFG Bioscience
New Product
BZY 2 4-DI-O-BENZOYL-Α-DMANNOPYRNS 500MG
Catalog # 77888-673
Supplier: AFG Bioscience
Supplier Number: C50750-0.5G
New Product
Frequently Bought Together
Eveready® Alkaline Batteries, Bulbtronics
Supplier: Bulbtronics
Polyethylene Disposable Apron
Supplier: Dival Safety & Supplies
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Supplier: MilliporeSigma
Ammonium persulfate (APS) ≥98.0% ACS analytical reagent
Supplier: VWR International
Bulk Syringes, Nonsterile, BD Medical
Supplier: BD
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Coagulase
Supplier: BD
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Perchloric acid standardised solution 0.1 N in acetic acid
Supplier: Thermo Scientific Chemicals
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
E.Z.N.A.® Plant & Fungal DNA Kit, Omega Bio-tek®
Supplier: Omega Bio-Tek
SupaMega Cassette, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris