DIGENEASIDE SODIUM SALT 10MG
Catalog # 77888-622
Supplier: AFG Bioscience
New Product
DIGENEASIDE SODIUM SALT 10MG
Catalog # 77888-622
Supplier: AFG Bioscience
Supplier Number: C50019-10MG
New Product
Frequently Bought Together
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Single-Use Needles, BD Medical
Supplier: BD
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Leakbuster™ Specimen Containers
Supplier: Starplex
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
n-Hexane ≥95%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Sandwich Bags, Low Density Flip Top Elkay Plastics
Supplier: Elkay Plastics
ACETIC RINSE SOLUTION 4L
Supplier: Ostrem Chemical
Human Recombinant FGF-basic (from E. coli)
Supplier: Peprotech
Succinimide 98+%
Supplier: Thermo Scientific Chemicals
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Dri-Contrad® Powder Detergent
Supplier: Decon Labs
Bovine Serum Albumin (BSA)
Supplier: Rockland Immunochemical
Corning® HBSS (Hanks Balanced Salt Solution), Corning
Supplier: Corning
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC