LACTO-N-TETRAOSE 25MG
Catalog # 77889-050
Supplier: AFG Bioscience
New Product
LACTO-N-TETRAOSE 25MG
Catalog # 77889-050
Supplier: AFG Bioscience
Supplier Number: C79024-25MG
New Product
Frequently Bought Together
Single-Use Needles, BD Medical
Supplier: BD
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Acetonitrile ≥99.8% for HPLC, Supelco®
Supplier: MilliporeSigma
Petroleum spirit, 35…60 °C, GR ACS, Supelco®
Supplier: MilliporeSigma
Urine Collection Tubes and Kits, BD Medical
Supplier: BD
tri-Sodium citrate dihydrate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Cypermethrin Single-Component Organic Standard, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International