2-FUCOSYLLACTOSE 100MG
Catalog # 77889-042
Supplier: AFG Bioscience
New Product
2-FUCOSYLLACTOSE 100MG
Catalog # 77889-042
Supplier: AFG Bioscience
Supplier Number: C78016-100MG
New Product
Frequently Bought Together
Polyethylene Disposable Apron
Supplier: Dival Safety & Supplies
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Hydrochloric acid (<lt/> 0.027 mol/l; <lt/> 0.027 N) 0.01 N analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
VWR® Ultra Pure Grade Phosphate Buffered Saline
Supplier: VWR
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Formalin 10%, neutral buffered, VWR®
Supplier: VWR International
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex
Reagent Sets for Colorimeters, Hach
Supplier: Hach
CytoFLEX Sheath Fluid, Beckman Coulter
Supplier: Beckman Coulter
AGAR BRAIN HEART INFUSION 450 G
Supplier: BECTON DICKINSON DE MEXICO, S. MX
Hydrazinium sulfate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Accessories for Laboratory Turbidimeters, Hach
Supplier: Hach
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Tissue Marking Dyes, Azer Scientific
Supplier: Azer Scientific
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics