4-METHOXYLPHENYL2-ACTAMD-2-DEOXY 1G
Catalog # 77888-945
Supplier: AFG Bioscience
New Product
4-METHOXYLPHENYL2-ACTAMD-2-DEOXY 1G
Catalog # 77888-945
Supplier: AFG Bioscience
Supplier Number: C53803-1G
New Product
Frequently Bought Together
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
Tissue Marking Dyes (TMD™), TBS®, General Data
Supplier: General Data Healthcare
Glycerine ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
VWR® Razor Blades
Supplier: VWR International
VWR® Reagent alcohol
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
Prinzing® Ball Valve Lockouts, Brady Worldwide®
Supplier: Brady Worldwide
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
E.Z.N.A.® Plasmid Mini Kit I, Omega Bio-tek®
Supplier: Omega Bio-Tek