2-ACTAMD-2-DEOXY-346-TRI-O 100MG
Catalog # 77888-942
Supplier: AFG Bioscience
New Product
2-ACTAMD-2-DEOXY-346-TRI-O 100MG
Catalog # 77888-942
Supplier: AFG Bioscience
Supplier Number: C53793-100MG
New Product
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Pocket Colorimeter II Test Kits
Supplier: Hach
VWR® Specimen Containers, PP with PE Caps
Supplier: VWR International
VWR® ClikSeal Specimen Containers
Supplier: VWR International
Gauze Pads, DUKAL™ Corporation
Supplier: Dukal
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Petroleum spirit, 35…60 °C, GR ACS, Supelco®
Supplier: MilliporeSigma
Acetone ACS
Supplier: INNOVA CHEMICALS CA
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BUFFER SOLN TOT CHLORINE 473ML
Supplier: Hach
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
CL17 DPD Indicator, Hach
Supplier: Hach
Chloroform ≥99.8% stabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics