6-AZIDO-6-DEOXY-L-FUCOSE PERACETATE 25MG
Catalog # 77888-751
Supplier: AFG Bioscience
New Product
6-AZIDO-6-DEOXY-L-FUCOSE PERACETATE 25MG
Catalog # 77888-751
Supplier: AFG Bioscience
Supplier Number: C51554-25MG
New Product
Frequently Bought Together
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Hydrogen peroxide 30%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®
Supplier: VWR International
Chloroform ≥99.8% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® TRIS Buffer, 1 M Solution, pH 8.0
Supplier: VWR
Direct-zol™ RNA Kits, Zymo Research
Supplier: Zymo Research
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Caffeine ≥99.7%
Supplier: Thermo Scientific Chemicals
L(-)-Proline 99%
Supplier: Thermo Scientific Chemicals
Calcein AM fluorescent dye, Corning®
Supplier: Corning
GASTIGHT® 1000 Series Syringes, Hamilton
Supplier: Hamilton
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
Small Red Biohazard Bags, Unimed-Midwest
Supplier: Unimed-Midwest
EDTA 0.800 M, cartridge for digital titrator
Supplier: Hach