2-ACETAMID-346-TRI-O-ACTYL2 25MG
Catalog # 77888-941
Supplier: AFG Bioscience
New Product
2-ACETAMID-346-TRI-O-ACTYL2 25MG
Catalog # 77888-941
Supplier: AFG Bioscience
Supplier Number: C53791-25MG
New Product
Frequently Bought Together
D-(+)-Sucrose, Ultrapure DNase-, RNase-free
Supplier: VWR
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Supplier: Thermo Fisher Scientific
Sodium hydroxide ≥97.0%, pellets ACS
Supplier: VWR
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Supplier: MilliporeSigma
Preserved Starfish
Supplier: Avantor
PEI MAX® Transfection Reagents (Linear Polyethylenimine Hydrochloride)
Supplier: KYFORA BIO LLC.
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Supplier: VWR International
Potassium hydroxide 0.100 N (N/10) in isopropanol (IPA)
Supplier: Ricca Chemical
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
L(+)-Tartaric acid 10% (w/v) in aqueous solution
Supplier: Ricca Chemical
BsoBI
Supplier: New England Biolabs (NEB)
Sodium Nitrite
Supplier: Ward's Science
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
Supplier: Thermo Scientific
Genomic DNA Clean & Concentrator™ Kits, Zymo Research
Supplier: Zymo Research