2-DEOXY-2-FLUORO-D-GALACTOSE 25MG
Catalog # 77888-889
Supplier: AFG Bioscience
New Product
2-DEOXY-2-FLUORO-D-GALACTOSE 25MG
Catalog # 77888-889
Supplier: AFG Bioscience
Supplier Number: C52519-25MG
New Product
Frequently Bought Together
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Alconox® Powdered Precision Cleaners
Supplier: Alconox
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
Cytology Funnels for Shandon CytoSpin™ Centrifuge, Biomedical Polymers
Supplier: Biomedical Polymers
Leakbuster™ Specimen Containers
Supplier: Starplex
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BPMO ≥99% (by 1H-NMR), high purity
Supplier: Enzo Life Sciences
Chloroform-[D₁] (99.8% D) (Isotopic) + 0.03% v/v TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
BBL™ SFP Agar Base, BD
Supplier: BD
Difco™ YPD Medium, BD Life Sciences
Supplier: BD
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International